accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOH

CAS RN 111-46-6

IUPAC Name: 2-(2-hydroxyethoxy)ethan-1-ol
Synonyms: diethylene glycol 2,2'-dihydroxydiethyl ether epa pesticide chemical code 338200 β,β'-dihydroxydiethyl ether DEG 2,2'-oxyethanol diethylenglykol brecolane ndg deactivator E diethylene glycol (deg)
Molecular Weight (g/mol): 106.12
Molecular Formula: C4H10O3
InChi Key: MTHSVFCYNBDYFN-UHFFFAOYSA-N
SMILES: OCCOCCO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (5)
  • (4)
  • (3)
Physical Form 
  • (9)
  • (3)
Percent Purity 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (4)
  • (3)
  • (9)
  • (3)
  • (3)
  • (7)
  • (1)
  • (3)
  • (1)
  • (1)