accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONHOH(Z)

CAS RN 111-58-0

IUPAC Name: (9Z)-N-(2-hydroxyethyl)octadec-9-enamide
Synonyms: oleoylethanolamide oleamide mea N-(hydroxyethyl) oleamide N-oleoyl ethanolamide
Molecular Weight (g/mol): 325.54
Molecular Formula: C20H39NO2
InChi Key: BOWVQLFMWHZBEF-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)NCCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results