missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Oleylethanolamide
GREENER_CHOICE

Product Code. 18075882
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075882 50 mg 50mg
18075872 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075882 Supplier Tocris Bioscience™ Supplier No. 1484/50
  • ONLINE EXCLUSIVE PRICE
    582.00€
    550.20€ / 50mg
    Save 31.80€
    5% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipid mediator, anorexic actions; also GPR55 and PPARα agonist

Lipid mediator and analog of anandamide (Cat. No. 1339) that is involved in peripheral regulation of feeding. Selective GPR55 agonist (EC50 values are 0.44, >30 and >30 μM at GPR55, CB1 and CB2 respectively) and PPARα agonist (EC50 = 120 nM). Induces satiety through activation of PPARα and is also a ceramidase inhibitor. Also endogenous agonist at the GPR119 receptor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-58-0
Molecular Formula C20H39NO2

Specifications

Quantity 50 mg
Formula Weight 325.53
Purity >99%
Molecular Weight (g/mol) 325.537
Chemical Name or Material Oleylethanolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.