accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOH

CAS RN 111-76-2

IUPAC Name: 2-butoxyethan-1-ol
Synonyms: butoxyethanol butyl cellosolve ethylene glycol monobutyl ether (egbe) butyl oxitol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: POAOYUHQDCAZBD-UHFFFAOYSA-N
SMILES: CCCCOCCO
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (12)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (4)
Physical Form 
  • (12)
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (12)
  • (2)
  • (3)
  • (3)
  • (4)
  • (12)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)