CAS RN 111544-93-5
IUPAC Name:
1,3-diethyl 2-(2,4-dichlorophenyl)propanedioate
Synonyms:
1,3-diethyl 2-(2,4-dichlorophenyl)propanedioate
Molecular Weight (g/mol):
305.15
Molecular Formula:
C13H14Cl2O4
InChi Key:
FIGCCTUCURBNIF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl
1
–
1
of
1
results
1
Diethyl (2,4-dichlorophenyl)malonate, 95%
CAS: 111544-93-5 Molecular Formula: C13H14Cl2O4 Molecular Weight (g/mol): 305.15 MDL Number: MFCD00831103 InChI Key: FIGCCTUCURBNIF-UHFFFAOYSA-N Synonym: diethyl 2-2,4-dichlorophenyl malonate,diethyl 2,4-dichlorophenyl malonate,1,3-diethyl 2-2,4-dichlorophenyl propanedioate,diethyl 2-2,4-dichlorophenyl propanedioate,diethyl 2,4-dichlorophenyl propanedioate,diethyl 2,4-dichlorophenylmalonate,2-2,4-dichloro-phenyl-malonic acid diethyl ester,propanedioic acid,2-2,4-dichlorophenyl-,1,3-diethyl ester PubChem CID: 2736112 IUPAC Name: diethyl 2-(2,4-dichlorophenyl)propanedioate SMILES: CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl