accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1119-44-4

CH3CH3O(E)

CAS RN 1119-44-4

IUPAC Name: (3E)-hept-3-en-2-one
Synonyms: (3E)-hept-3-en-2-one
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: JHHZQADGLDKIPM-AATRIKPKSA-N
SMILES: CCC\C=C\C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results