accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOH

CAS RN 112-25-4

IUPAC Name: 2-(hexyloxy)ethan-1-ol
Synonyms: hexyloxyethanol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: UPGSWASWQBLSKZ-UHFFFAOYSA-N
SMILES: CCCCCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results