accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOH

CAS RN 112-34-5

IUPAC Name: 2-(2-butoxyethoxy)ethan-1-ol
Synonyms: butoxydiglycol diethylene glycol monobutyl ether (degbe) deg monobutyl ether
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: OAYXUHPQHDHDDZ-UHFFFAOYSA-N
SMILES: CCCCOCCOCCO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)