accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-73-2

H3COOOCH3

CAS RN 112-73-2

IUPAC Name: 1-[2-(2-butoxyethoxy)ethoxy]butane
Synonyms: diethylene glycol dibutyl ether
Molecular Weight (g/mol): 218.34
Molecular Formula: C12H26O3
InChi Key: KZVBBTZJMSWGTK-UHFFFAOYSA-N
SMILES: CCCCOCCOCCOCCCC
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)