accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1121-60-4

IUPAC Name: pyridine-2-carbaldehyde
Synonyms: 2-formylpyridine
Molecular Weight (g/mol): 107.11
Molecular Formula: C6H5NO
InChi Key: CSDSSGBPEUDDEE-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)