CAS RN 112111-44-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(+)-Modafinic Acid, TRC
CAS: 112111-44-1 Molecular Formula: C15 H14 O3 S Molecular Weight (g/mol): 274.33 Synonym: Acetic acid, 2-[(S)-(diphenylmethyl)sulfinyl]-,Acetic acid, [(S)-(diphenylmethyl)sulfinyl]- (9CI),2-[(S)-(Diphenylmethyl)sulfinyl]acetic acid,(S)-(+)-Modafinil acid,[(S)-(Diphenylmethyl)sulfinyl]acetic acid IUPAC Name: 2-[(S)-benzhydrylsulfinyl]acetic acid SMILES: OC(=O)C[S@](=O)C(c1ccccc1)c2ccccc2