accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SCN

CAS RN 1122-82-3

IUPAC Name: isothiocyanatocyclohexane
Synonyms: cyclohexane, isothiocyanato-
Molecular Weight (g/mol): 141.23
Molecular Formula: C7H11NS
InChi Key: MZSJGCPBOVTKHR-UHFFFAOYSA-N
SMILES: S=C=NC1CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)