missing translation for 'onlineSavingsMsg'
Learn More

Isothiocyanatocyclohexane, TRC

Código de producto. 30397509
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397509 Supplier Toronto Research Chemicals Supplier No. TRCI9051151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isothiocyanatocyclohexane
CAS 1122-82-3
Molecular Formula C7 H11 N S
Quantity 1 g
Synonym Isothiocyanic Acid Cyclohexyl Ester, Cyclohexyl Isothiocyanate, NSC 60129
InChI Formula InChI=1S/C7H11NS/c9-6-8-7-4-2-1-3-5-7/h7H,1-5H2
SMILES S=C=NC1CCCCC1
IUPAC Name isothiocyanatocyclohexane
Molecular Weight (g/mol) 141.23
Formula Weight 141.0612
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.