accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1123-00-8

OOH

CAS RN 1123-00-8

IUPAC Name: 2-cyclopentylacetic acid
Synonyms: cyclopentaneacetic acid
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: YVHAIVPPUIZFBA-UHFFFAOYSA-N
SMILES: OC(=O)CC1CCCC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)