CAS RN 112306-54-4
CAS RN 112306-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1-Dimethylbutylamine Hydrochloride, TRC
CAS: 112306-54-4 Molecular Formula: C6H16ClN Molecular Weight (g/mol): 137.65 Synonym: 2-Methyl-2-pentanamine Hydrochloride,NSC 99269 Hydrochloride SMILES: NC(C)(C)CCC.Cl