missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbutylamine Hydrochloride, TRC

Product Code. 30138662
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138662 1 g 1g
30010075 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138662 Supplier Toronto Research Chemicals Supplier No. TRCD4728251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112306-54-4
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
Synonym 2-Methyl-2-pentanamine Hydrochloride, NSC 99269 Hydrochloride
SMILES NC(C)(C)CCC.Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H15N.ClH/c1-4-5-6(2,3)7;/h4-5,7H2,1-3H3;1H
Formula Weight 137.0971
Chemical Name or Material 1,1-Dimethylbutylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.