accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONH2ClONHNOFHOOOHOHOOHO

CAS RN 112885-42-4

IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid; 4-amino-5-chloro-2-ethoxy-N-({4-[(4-fluorophenyl)methyl]morpholin-2-yl}methyl)benzamide
Synonyms: citric acid; mosapride
Molecular Weight (g/mol): 614.02
Molecular Formula: C27H33ClFN3O10
InChi Key: HUZTYZBFZKRPFG-UHFFFAOYNA-N
SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CCOC1=CC(N)=C(Cl)C=C1C(=O)NCC1CN(CC2=CC=C(F)C=C2)CCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results