accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOFF(E)

CAS RN 112897-97-9

IUPAC Name: (2E)-3-(3,4-difluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(3,4-difluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 183.13
Molecular Formula: C9H5F2O2
InChi Key: HXBOHZQZTWAEHJ-DUXPYHPUSA-M
SMILES: [O-]C(=O)\C=C\C1=CC=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results