accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1129-30-2

CH3ONCH3O

CAS RN 1129-30-2

IUPAC Name: 1-(6-acetylpyridin-2-yl)ethan-1-one
Synonyms: 2,6-diacetylpyridine
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: BEZVGIHGZPLGBL-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(=N1)C(C)=O
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)