accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3COOO

CAS RN 113-24-6

IUPAC Name: sodium 2-oxopropanoate
Synonyms: sodium pyruvate natriumpyruvat pyruvic acid, sodium salt
Molecular Weight (g/mol): 110.04
Molecular Formula: C3H3NaO3
InChi Key: DAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES: [Na+].CC(=O)C([O-])=O
Quantity 
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (7)
  • (8)
Physical Form 
  • (5)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (3)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (7)
  • (8)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)