missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic acid Na-salt, Serva Electrophoresis™

Product Code. 11761877
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11761877 25 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11761877 Supplier Serva Electrophoresis Supplier No. 1522001
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 113-24-6
Color White
Loss on Drying max. 0.5 %
Molecular Formula C3H3NaO3
Quantity 25 g
Synonym sodium pyruvate, sodium 2-oxopropanoate, pyruvic acid sodium salt, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
Solubility Information 1.5g in 25mL H2O
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Molecular Weight (g/mol) 110.04
PubChem CID 23662274
ChEBI CHEBI:50144
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.