accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KH3CNCH3NSClCH3CH3S(R)(R)NHOON(S)OO

CAS RN 113-98-4

IUPAC Name: potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: potassium benzylpenicillin(1-) chlorpromazine
Molecular Weight (g/mol): 691.34
Molecular Formula: C33H36ClKN4O4S2
InChi Key: HTQQVOYPSGNVPA-HBRCYENSSA-M
SMILES: [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (10)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (7)
  • (1)
Color 
  • (7)
Type 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (4)
  • (3)
  • (3)
  • (10)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (7)
  • (1)
  • (7)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)