missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Penicillinate Potassium Salt, TRC

Product Code. 30137441
Change view
Click to view available options
Quantity:
250 mg
5 g
50 g
Unit Size:
250mg
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137441 5 g 5g
30137440 50 g 50g
17914555 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30137441 Supplier Toronto Research Chemicals Supplier No. TRCB2886015G
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-98-4
Synonym Benzylpenicillin Potassium∣(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid potassium salt (1:1)∣3,3-Dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt∣(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt∣[2S-(2α,5α,6β)]-3,3-Dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monopotassium salt∣Penicillin G potassium salt∣Benzylpenicillin potassium∣Benzylpenicillin potassium salt∣Benzylpenicillinic acid potassium salt∣Cosmopen∣Cristapen∣Crytapen∣Eskacillin∣Falapen∣Forpen∣Hipercilina∣Hyasorb∣Hylenta∣M-Cillin∣Megacillin tablets∣Monopen∣NSC 131815∣Notaral∣Novocillin vet.∣Penicillin G potassium∣Pentid∣Pentids∣Potassium 2,2-dimethyl-6β-phenylacetamidopenam-3α-carboxylate∣Potassium 6-(phenylacetamido)penicillanate∣Potassium benzylpenicillin∣Potassium benzylpenicillinate∣Potassium penicillin G∣Scotcil∣Tabilin

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 372.48
Product Type Metabolite∣API
Quantity 5 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.