accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 114-80-7

BrCH3NH3COONH3CH3CCH3

CAS RN 114-80-7

IUPAC Name: 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide
Synonyms: neostigmine bromide neostigmine bromide [inn:ban:jan]
Molecular Weight (g/mol): 303.2
Molecular Formula: C12H19BrN2O2
InChi Key: LULNWZDBKTWDGK-UHFFFAOYSA-M
SMILES: [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)