missing translation for 'onlineSavingsMsg'
Learn More

Neostigmine Bromide, TRC

Product Code. 30477101
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477101 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477101 Supplier Toronto Research Chemicals Supplier No. TRCN39001025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neostigmine Bromide
CAS 114-80-7
Molecular Formula C12 H19 N2 O2 . Br
Quantity 25 mg
Synonym Benzenaminium','3-[[(dimethylamino)carbonyl]oxy]-N,N,N-trimethyl-','bromide, (m-Hydroxyphenyl)trimethylammonium bromide','dimethylcarbamate, Ammonium','(m-hydroxyphenyl)trimethyl-','bromide','dimethylcarbamate, Benzenaminium','3-[[(dimethylamino)carbonyl]oxy]-N,N,N-trimethyl-','bromide, Neostigmine bromide, Carbamic acid','dimethyl-','ester with (m-hydroxyphenyl)trimethylammonium bromide, (3-Dimethylcarbamoyloxyphenyl)trimethylammonium bromide, Eustigmin bromide, Kirkstigmine bromide, Neoeserine bromide, Neoesserin, Philostigmin bromide, Prostigmin bromide, Prostigmine bromide, Stigmanol bromide, Stigmosan bromide, Syntostigmin (tablet), Syntostigmin bromide, Vagostigmine bromide
InChI Formula InChI=1S/C12H19N2O2.BrH/c1-13(2)12(15)16-11-8-6-7-10(9-11)14(3,4)5;/h6-9H,1-5H3;1H/q+1;/p-1
SMILES [Br-].CN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C
IUPAC Name [3-(dimethylcarbamoyloxy)phenyl]-trimethylazanium;bromide
Molecular Weight (g/mol) 303.2
Formula Weight 302.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.