accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONHNN(Z)

CAS RN 1141-59-9

IUPAC Name: (4Z)-3-hydroxy-4-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: (4Z)-3-hydroxy-4-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 215.21
Molecular Formula: C11H9N3O2
InChi Key: VLCAILLZPUINNF-LCYFTJDESA-N
SMILES: OC1=CC(=O)C=C\C1=N\NC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (8)
  • (7)
Quantity 
  • (6)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
  • (2)
Physical Form 
  • (5)
  • (1)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (4)
  • (3)
  • (1)
  • (8)
  • (7)
  • (6)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)