CAS RN 1143-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Urolithin A
PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC)
2
Urolithin A, TRC
CAS: 1143-70-0 Molecular Formula: C13 H8 O4 Molecular Weight (g/mol): 228.2 Synonym: 6H-Dibenzo[b,d]pyran-6-one, 3,8-dihydroxy- (9CI, ACI),2-Biphenylcarboxylic acid, 2',4,4'-trihydroxy-, δ-lactone (7CI, 8CI),3,8-Dihydroxy-6H-dibenzo[b,d]pyran-6-one (ACI),2',7-Dihydroxy-3,4-benzocoumarin,3,8-Dihydroxybenzo[c]chromen-6-one,3,8-Dihydroxyurolithin,3,8-Hydroxydibenzo-α-pyrone,Urolithin A IUPAC Name: 3,8-dihydroxybenzo[c]chromen-6-one SMILES: Oc1ccc2c(OC(=O)c3cc(O)ccc23)c1