missing translation for 'onlineSavingsMsg'
Learn More

Urolithin A, TRC

Product Code. 30009263
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009263 5 mg 5mg
30006918 10 mg 10mg
30007143 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009263 Supplier Toronto Research Chemicals Supplier No. TRCU8470005MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1143-70-0
Molecular Formula C13 H8 O4
Molecular Weight (g/mol) 228.2
Synonym 6H-Dibenzo[b,d]pyran-6-one, 3,8-dihydroxy- (9CI, ACI), 2-Biphenylcarboxylic acid, 2',4,4'-trihydroxy-, δ-lactone (7CI, 8CI), 3,8-Dihydroxy-6H-dibenzo[b,d]pyran-6-one (ACI), 2',7-Dihydroxy-3,4-benzocoumarin, 3,8-Dihydroxybenzo[c]chromen-6-one, 3,8-Dihydroxyurolithin, 3,8-Hydroxydibenzo-α-pyrone, Urolithin A
IUPAC Name 3,8-dihydroxybenzo[c]chromen-6-one
SMILES Oc1ccc2c(OC(=O)c3cc(O)ccc23)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H8O4/c14-7-1-3-9-10-4-2-8(15)6-12(10)17-13(16)11(9)5-7/h1-6,14-15H
Formula Weight 228.0423
Chemical Name or Material Urolithin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.