accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3CH3HO

CAS RN 115-22-0

IUPAC Name: 3-hydroxy-3-methylbutan-2-one
Synonyms: 3-hydroxy-3-methyl-2-butanone
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: BNDRWEVUODOUDW-UHFFFAOYSA-N
SMILES: CC(=O)C(C)(C)O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results