accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOOHO

CAS RN 117-12-4

IUPAC Name: 1,5-dihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: anthrarufin Az-U
Molecular Weight (g/mol): 240.21
Molecular Formula: C14H8O4
InChi Key: JPICKYUTICNNNJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)C1=CC=CC(O)=C1C2=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results