accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 117-84-0

IUPAC Name: 1,2-dioctyl benzene-1,2-dicarboxylate
Synonyms: dioctyl phthalate DNOP vinicizer 85 dinopol nop dioktylester kyseliny ftalove dicapryl phthalate polycizer 162 rcra waste number U107
Molecular Weight (g/mol): 390.56
Molecular Formula: C24H38O4
InChi Key: MQIUGAXCHLFZKX-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)