accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OHH3CH3CCH3CH3H3CCH3HOH3CH3CCH3

CAS RN 118-82-1

IUPAC Name: 2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phenol
Synonyms: ionox 220 MB 1 (antioxidant) ionox 220 antioxidant binox M antioxidant E 702 ethyl 702 bimox M LZ-MB 1
Molecular Weight (g/mol): 424.67
Molecular Formula: C29H44O2
InChi Key: MDWVSAYEQPLWMX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(CC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results