accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 118427-29-5

IUPAC Name: [4-(propan-2-yl)phenyl]hydrazine
Synonyms: (4-isopropylphenyl)hydrazine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: ZYATZFJUOXJFPY-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(NN)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results