missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenylhydrazine hydrochloride, 98%, Thermo Scientific™

Product Code. 10175940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10175940 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10175940 Supplier Thermo Scientific Acros Supplier No. 306150050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 118427-29-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 217°C
Color Beige to White or Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H14N2
Linear Formula (CH3)2CHC6H4NHNH2·HCl
MDL Number MFCD00013461
Quantity 5 g
Synonym 4-isopropylphenylhydrazine hydrochloride, 4-isopropylphenyl hydrazine hydrochloride, 1-4-isopropylphenyl hydrazine hydrochloride, 4-propan-2-yl phenyl hydrazine hydrochloride, hydrazine, 4-1-methylethyl phenyl-, monohydrochloride, 4-isopropylphenylhydrazinehydrochloride, pubchem7533, acmc-1cbwc, 4-propan-2-ylphenyl hydrazine hydrochloride, ksc183e0j
InChI Key ZYATZFJUOXJFPY-UHFFFAOYSA-N
SMILES CC(C)C1=CC=C(NN)C=C1
IUPAC Name (4-propan-2-ylphenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 150.23
PubChem CID 2733248
Formula Weight 186.68
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material 4-Isopropylphenylhydrazine hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.