accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFBr

CAS RN 1186194-80-8

IUPAC Name: 1-bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
Synonyms: 1-bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: VDKXBQCWYHXJNB-UHFFFAOYSA-N
SMILES: FC1=CC(CC(F)(F)F)=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results