missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene, 95%, Thermo Scientific™

Product Code. 15418455
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418455 250 mg 250mg
15408455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418455 Supplier Thermo Scientific Alfa Aesar Supplier No. H33404.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1186194-80-8
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.026
MDL Number MFCD11226691
InChI Key VDKXBQCWYHXJNB-UHFFFAOYSA-N
Synonym 1-bromo-4-fluoro-2-2,2,2-trifluoroethyl benzene
PubChem CID 45790615
IUPAC Name 1-bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
SMILES C1=CC(=C(C=C1F)CC(F)(F)F)Br

Specifications

Boiling Point 76°C (15 mmHg)
Quantity 250 mg
Formula Weight 257.02
Percent Purity 95%
Chemical Name or Material 1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.