accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1189-09-9

CH3OOH3CH3CH3C(E)

CAS RN 1189-09-9

IUPAC Name: methyl (2E)-3,7-dimethylocta-2,6-dienoate
Synonyms: methyl dimethyloctadienoate
Molecular Weight (g/mol): 182.26
Molecular Formula: C11H18O2
InChi Key: ACOBBFVLNKYODD-CSKARUKUSA-N
SMILES: COC(=O)\C=C(/C)CCC=C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results