accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119-67-5

HOOO

CAS RN 119-67-5

IUPAC Name: 2-formylbenzoic acid
Synonyms: 2-carboxybenzaldehyde phthalaldehydic acid
Molecular Weight (g/mol): 150.13
Molecular Formula: C8H6O3
InChi Key: DYNFCHNNOHNJFG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)