accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2N(S)

CAS RN 119237-64-8

IUPAC Name: (2S)-2-(diphenylmethyl)pyrrolidin-1-ium
Synonyms: 2-diphenylmethylpyrrolidine
Molecular Weight (g/mol): 238.35
Molecular Formula: C17H20N
InChi Key: OXOBKZZXZVFOBB-INIZCTEOSA-O
SMILES: C1C[NH2+][C@@H](C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results