CAS RN 1192559-25-3
IUPAC Name:
(5-fluoropyridin-2-yl)thiourea
Synonyms:
5-fluoropyridin-2-ylthiourea
Molecular Weight (g/mol):
171.19
Molecular Formula:
C6H6FN3S
InChi Key:
MAMMTYFIMUFYQT-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=NC=C(F)C=C1
1
–
1
of
1
results
1
N-(5-Fluoro-2-pyridyl)thiourea, 97%, Thermo Scientific™
CAS: 1192559-25-3 Molecular Formula: C6H6FN3S Molecular Weight (g/mol): 171.193 MDL Number: MFCD17780187 InChI Key: MAMMTYFIMUFYQT-UHFFFAOYSA-N Synonym: 1-5-fluoropyridin-2-yl thiourea,thiourea, n-5-fluoro-2-pyridinyl,n-5-fluoro-2-pyridyl thiourea,n-5-fluoropyridin-2-yl thiourea PubChem CID: 52940344 IUPAC Name: (5-fluoropyridin-2-yl)thiourea SMILES: C1=CC(=NC=C1F)NC(=S)N