accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 119446-68-3

IUPAC Name: 1-({2-[2-chloro-4-(4-chlorophenoxy)phenyl]-4-methyl-1,3-dioxolan-2-yl}methyl)-1H-1,2,4-triazole
Synonyms: difenoconazole
Molecular Weight (g/mol): 406.26
Molecular Formula: C19H17Cl2N3O3
InChi Key: BQYJATMQXGBDHF-UHFFFAOYNA-N
SMILES: CC1COC(CN2C=NC=N2)(O1)C1=CC=C(OC2=CC=C(Cl)C=C2)C=C1Cl
Molecular Weight (g/mol) 
  • (7)
  • (4)
Concentration 
  • (2)
  • (2)
  • (2)
Solvent or Matrix 
  • (1)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (7)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)