missing translation for 'onlineSavingsMsg'
Learn More

Difenoconazol, Fluka™

Product Code. 16202912 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16202912 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16202912 Supplier Honeywell-Fluka Supplier No. R1624100MG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Analytical standard, reference material

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Difenoconazol
Packaging Glass Bottle
CAS 119446-68-3
Color White
Melting Point 82°C
Synonym 1-[(2-[2-Chloro-4-(4-chloro-phenoxy)-phenyl]-4-methyl[1,3]dioxolan-2-yl)methyl]-1H-1,2,4-triazole
Industry Type Environmental Analysis
Molecular Weight (g/mol) 406.26
Quantity 100 mg
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.