accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHN

CAS RN 1195-59-1

IUPAC Name: [6-(hydroxymethyl)pyridin-2-yl]methanol
Synonyms: [6-(hydroxymethyl)pyridin-2-yl]methanol
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: WWFMINHWJYHXHF-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(CO)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results