accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O(S)(S)H3CH3C

CAS RN 1196-01-6

IUPAC Name: (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-one
Synonyms: levoverbenone
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: DCSCXTJOXBUFGB-JGVFFNPUSA-N
SMILES: CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results