missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TerpiMix 1 2500 μg/mL in Isopropanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | (-)-Verbenone∣(+)-Borneol∣(+)-Fenchone∣(+)-Fenchyl Alcohol∣(1S)-(+)-3-Carene∣1-Methyl-2-isopropylbenzene∣4-Cymene∣4-Thujanol∣Acetic Acid-Octyl Ester∣alpha-(-)-Thujone∣alpha-Terpinene∣alpha-Terpineol∣beta-Ocimene∣beta-Pinene∣Camphene∣DL-alpha-Pinene∣DL-Linalool∣Eucalyptol (1,8-Cineole)∣gamma-Terpinene∣Myrcene∣Sabinene∣Terpinolene |
| CAS | 1196-01-6, 464-43-7, 4695-62-9, 2-9-2217, 498-15-7, 527-84-4 |
| Analyte or Component Names | (-)-Verbenone∣(+)-Borneol∣(+)-Fenchone∣(+)-Fenchyl Alcohol∣(1S)-(+)-3-Carene∣1-Methyl-2-isopropylbenzene∣4-Cymene∣4-Thujanol∣Acetic Acid-Octyl Ester∣alpha-(-)-Thujone∣alpha-Terpinene∣alpha-Terpineol∣beta-Ocimene∣beta-Pinene∣Camphene∣DL-alpha-Pinene∣DL-Linalool∣Eucalyptol (1,8-Cineole)∣gamma-Terpinene∣Myrcene∣Sabinene∣Terpinolene |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Isopropanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | (-)-Verbenone 2500 μg/mL∣(+)-Borneol 2500 μg/mL∣(+)-Fenchone 2500 μg/mL∣(+)-Fenchyl Alcohol 2500 μg/mL∣(1S)-(+)-3-Carene 2500 μg/mL∣1-Methyl-2-isopropylbenzene 2500 μg/mL∣4-Cymene 2500 μg/mL∣4-Thujanol 2500 μg/mL∣Acetic Acid-Octyl Ester 2500 μg/mL∣alpha-(-)-Thujone 2500 μg/mL∣alpha-Terpinene 2500 μg/mL∣alpha-Terpineol 2500 μg/mL∣beta-Ocimene 2500 μg/mL∣beta-Pinene 2500 μg/mL∣Camphene 2500 μg/mL∣DL-alpha-Pinene 2500 μg/mL∣DL-Linalool 2500 μg/mL∣Eucalyptol (1,8-Cineole) 2500 μg/mL∣gamma-Terpinene 2500 μg/mL∣Myrcene 2500 μg/mL∣Sabinene 2500 μg/mL∣Terpinolene 2500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?