CAS RN 120-82-1
1,2,4-Trichlorobenzene, 99%
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
1,2,4-Trichlorobenzene, 99%, pure
CAS: 120-82-1 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
HJ 350-2007 SVOC Mixture 620 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
SVOC Mixture 1003 1000 μg/mL in Benzene:Dichloromethane (3:1), Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
EPA Method 625 SV Calibration Mixture 660 1000 μg/mL in Benzene:Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
EPA Method 8270 UltiMix Calibration Mixture 500-1000 μg/mL in Benzene:Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
EPA Method 8270 Calibration Mixture 500-1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
CLP SVOC Calibration Mixture 512 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
EPA Method 8270 LCS Mixture 100 μg/mL in Acetone:MeCl2 83.5:16.5, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
1,2,4-Trichlorobenzene 5000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
1,2,4-Trichlorobenzene, ReagentPlus™, ≥99%, Solstice
CAS: 120-82-1 Molecular Formula: C6H3Cl3 Molecular Weight (g/mol): 181.44 MDL Number: MFCD00000547 InChI Key: PBKONEOXTCPAFI-UHFFFAOYSA-N Synonym: benzene, 1,2,4-trichloro,unsym-trichlorobenzene,hostetex l-pec,trojchlorobenzen,1,2,4-trichlorbenzol,1,2,4-trichlorobenzol,1,3,4-trichlorobenzene,trichlorobenzene a,1,2,5-trichlorobenzene,as-trichlorobenzene PubChem CID: 13 ChEBI: CHEBI:28222 IUPAC Name: 1,2,4-trichlorobenzene SMILES: C1=CC(=C(C=C1Cl)Cl)Cl
HJ 834-2017, HJ 951-2018 VOC & SVOCs Mixture 155 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
- 1
- 2