missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichlorobenzene, TRC

Product Code. 30456095
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456095 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30456095 Supplier Toronto Research Chemicals Supplier No. TRCT77422010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Trichlorobenzene
CAS 120-82-1
Molecular Formula C6 H3 Cl3
Quantity 10 g
Synonym Benzene','1,2,4-trichloro-, 1,2,4-Trichlorobenzene, 1,2,4-Trichlorobenzol, 1,2,5-Trichlorobenzene, 1,3,4-Trichlorobenzene, Hostetex L-PEC, NSC 406697, unsym-Trichlorobenzene
InChI Formula InChI=1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H
SMILES Clc1ccc(Cl)c(Cl)c1
IUPAC Name 1,2,4-trichlorobenzene
Molecular Weight (g/mol) 181.45
Formula Weight 179.93
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.