CAS RN 120013-84-5
CAS RN 120013-84-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
rac-(cis/trans) Donepezil N-Oxide, TRC
CAS: 120013-84-5 Molecular Formula: C24 H29 N O4 Molecular Weight (g/mol): 395.49 Synonym: Donepezil N-Oxide IUPAC Name: 2-[(1-benzyl-1-oxidopiperidin-1-ium-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one SMILES: COc1cc2CC(CC3CC[N+]([O-])(Cc4ccccc4)CC3)C(=O)c2cc1OC
2
Donepezil N-Oxide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.