CAS RN 1206593-22-7
IUPAC Name:
4-ethoxy-3-(trifluoromethyl)benzonitrile
Synonyms:
4-ethoxy-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
215.18
Molecular Formula:
C10H8F3NO
InChi Key:
GQHVMIHFKJWGPT-UHFFFAOYSA-N
SMILES:
CCOC1=C(C=C(C=C1)C#N)C(F)(F)F
1
–
1
of
1
results
1
4-Ethoxy-3-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™
CAS: 1206593-22-7 Molecular Formula: C10H8F3NO Molecular Weight (g/mol): 215.18 MDL Number: MFCD13196579 InChI Key: GQHVMIHFKJWGPT-UHFFFAOYSA-N Synonym: 4-ethoxy-3-trifluoromethyl benzonitrile,4-ethoxy-3-trifluoromethyl benzenecarbonitrile PubChem CID: 57361402 IUPAC Name: 4-ethoxy-3-(trifluoromethyl)benzonitrile SMILES: CCOC1=C(C=C(C=C1)C#N)C(F)(F)F