CAS RN 12068-69-8
IUPAC Name:
dibismuth(3+) triselandiide
Synonyms:
bismuth selenide
Molecular Weight (g/mol):
654.87
Molecular Formula:
Bi2Se3
InChi Key:
FBGGJHZVZAAUKJ-UHFFFAOYSA-N
SMILES:
[Se--].[Se--].[Se--].[Bi+3].[Bi+3]
1
–
2
of
2
results
1
Bismuth(III) selenide, 99.995% (metals basis), Thermo Scientific™
CAS: 12068-69-8 MDL Number: MFCD00014200 Synonym: dibismuth selenium 2-,dibismuth triselenide,dibismuth 3+ ion triselenide,bismuth selenide,,bismuth iii selenide trace metals basis 10g
2
Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis)
CAS: 12068-69-8 MDL Number: MFCD00014200 Synonym: dibismuth selenium 2-,dibismuth triselenide,dibismuth 3+ ion triselenide,bismuth selenide,,bismuth iii selenide trace metals basis 10g